Learn More
Sino Biological Human FUT10 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11327T04100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FUT10 (rh FUT10; Catalog#11327-H07H; Q6P4F1-1; Leu32-Asp479). FUT10 specific IgG was purified by Human FUT10 affinity chromatography.
Catalog No. 50-237-0490