Learn More
Sino Biological Anti-SSEA1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 310171MM06100
Encompass_Preferred
Produced in mouse immunized with E. coli-derived Human SSEA1 / FUT4 fragment. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1110