Learn More
BIOHIPPO Human FXR Microsample
SDP
Catalog No. 5000313785
Encompass
Human FXR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FXR in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FXR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FXR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FXR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13785 Supplier BIOHIPPO Supplier No. BHE15211910
Human FXR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FXR in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FXR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FXR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FXR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color