Learn More
Raybiotech Inc Human GCSF R (CD114) ELISA Kit
Supplier:  Raybiotech Inc ELHGCSFR1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human G-CSF R/CD114.
Catalog No. 50-194-9302