Learn More
Biomatik Corporation Human G Protein Alpha 11 (GNa11) ELISA Kit
Supplier:  Biomatik Corporation EKU04294
Encompass_Preferred
Human G Protein Alpha 11 (GNa11) ELISA Kit for detecting GNa11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29992. Gene ID: 2767. Target Synonyms: GA11; Guanine Nucleotide-Binding Protein Subunit Alpha-11; Guanine nucleotide-binding protein G (y) subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8970