Learn More
Biomatik Corporation Human G Protein Alpha Transducing Activity Polypeptide 1 (GNaT1) ELISA Kit
Supplier:  Biomatik Corporation EKU12070
Encompass_Preferred
Human G Protein Alpha Transducing Activity Polypeptide 1 (GNaT1) ELISA Kit for detecting GNaT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11488. Gene ID: 2779. Target Synonyms: GBT1; GNATR; Guanine Nucleotide Binding Protein Alpha Transducing Activity Polypeptide 1; Transducin alpha-1 chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5108