Learn More
Biomatik Corporation Human GAA (Lysosomal alpha-glucosidase) ELISA Kit
Supplier:  Biomatik Corporation EKF58762
Encompass_Preferred
Human GAA (Lysosomal alpha-glucosidase) ELISA Kit for detecting GAA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10253. Target Synonyms: GAA; Lysosomal alpha-Glucosidase; Acid alpha-Glucosidase; Acid Maltase; LYAG; Acid maltase; Aglucosidase alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6219