Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GABRG3 (aa 17-59) Control Fragment Recombinant Protein

Catalog No. RP101533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101533 Supplier Invitrogen™ Supplier No. RP101533
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111385. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GABRG3 is a family of proteins involved in the GABAergic neurotransmission of the mammalian central nervous system. GABRG3 is a member of the GABA-A receptor gene family of heteromeric pentameric ligand-gated ion channels through which GABA, the major inhibitory neurotransmitter in the mammalian brain, acts. GABA-A receptors are the site of action of a number of important pharmacologic agents including barbiturates, benzodiazepines, and ethanol (summary by Whiting et al., 1999). For additional general information about the GABA-A receptor gene family, see GABRA1 (MIM 137160).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99928
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 2567
Name Human GABRG3 (aa 17-59) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias B230362M20Rik; CAE2; ECA2; GABA(A) receptor subunit gamma-3; GABA(G) receptor; GABA(G) receptor, gamma 3; GABA-alpha receptor gamma-3 subunit; GABRG3; Gabrg-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, gamma 3; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 3; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 3; gamma-aminobutyric acid A receptor, gamma 3; gamma-aminobutyric acid receptor subunit gamma-3; gamma-aminobutyric acid type A receptor gamma3 subunit; GEFSP3
Common Name GABRG3
Gene Symbol GABRG3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ARSRKVEEDEYEDSSSNQKWVLAPKSQDTDVTLILNKLLREYD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.