Learn More
BIORTUS USA INC Human GAK Protein, 100UG
Catalog No. NC2953340
Encompass_Preferred
Gene Name: GAK. Uniprot: O14976. Construct:GST-TEV-GAK(14-351). Expression Host: E. coli. Purity: 85%.
Catalog No. NC2953340 Supplier BIORTUS USA INC Supplier No. BP0398402A
Gene Name: GAK. Uniprot: O14976. Construct:GST-TEV-GAK(14-351). Expression Host: E. coli. Purity: 85%.