Learn More
BIOHIPPO Human GAL12 Microsample
SDP
Catalog No. 5000314667
Encompass
Human GAL12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL12 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GAL12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GAL12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GAL12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14667 Supplier BIOHIPPO Supplier No. BHE15211301
Human GAL12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL12 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GAL12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GAL12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GAL12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color