Learn More
American Research Products Inc Human Galactose 1 phosphate ur
Catalog No. 5000327653
Encompass
Human Galactose-1-phosphate uridylyltransferase (GALT) ELISA kit from CUSABIO Detection Range 2 74 pg/ml-2000 pg/ml Sensitivity 2 74 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27653 Supplier American Research Products Inc Supplier No. CSBEL009226HU
Human Galactose-1-phosphate uridylyltransferase (GALT) ELISA kit from CUSABIO Detection Range 2 74 pg/ml-2000 pg/ml Sensitivity 2 74 pg/ml Sample serum plasma tissue homogenates Method Sandwich