Learn More
Biomatik Corporation Human GALC (Galactocerebrosidase) ELISA Kit
Supplier:  Biomatik Corporation EKF58648
Encompass_Preferred
Human GALC (Galactocerebrosidase) ELISA Kit for detecting GALC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P54803. Target Synonyms: GALC; Galactocerebrosidase; Galactocerebroside beta-galactosidase; Galactosylceraminidase; galactosylceramidase; Krabbe disease; Galactosylceramide beta-galactosidase; GALCERase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5439