Learn More
Biomatik Corporation Human Galectin 1 (GAL1) CLIA Kit
Supplier:  Biomatik Corporation EKU09914
Encompass_Preferred
Human Galectin 1 (GAL1) CLIA Kit for detecting GAL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09382. Gene ID: 3956. Target Synonyms: LGALS1; GBP; HLBP14; Galaptin; Lectin,Galactoside-Binding Soluble 1; 14 kDa laminin-binding protein; Lactose-binding lectin 1; Putative MAPK-activating protein PM12; S-Lac lectin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5159