Learn More
ACROBiosystems ACROBiosystems Human Galectin-1 / LGALS1 Protein, Tag Free (MALS verified)
Catalog No. 502106461
Encompass_Preferred
Recombinant Protein;100UG;Human Galectin-1, Tag Free (GA1-H4113) is expressed from E.coli cells. It contains AA Met 1 - Asp 135 (Accession # P09382-1).;This protein carries no "tag". The protein has a calculated MW of 14.71 kDa. The protein migrates as 13-15 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-6461 Supplier ACROBiosystems Supplier No. GA1H4113100UG
Recombinant Protein;100UG;Human Galectin-1, Tag Free (GA1-H4113) is expressed from E.coli cells. It contains AA Met 1 - Asp 135 (Accession # P09382-1).;This protein carries no "tag". The protein has a calculated MW of 14.71 kDa. The protein migrates as 13-15 kDa under reducing (R) condition (SDS-PAGE).