Learn More
ACROBiosystems ACROBiosystems Human Galectin-4 / LGALS4 / L36LBP Protein, His tag
Supplier:  ACROBiosystems GA4H5246100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Galectin-4, His Tag (GA4-H5246) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 323 (Accession # P56470-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 38.1 kDa. The protein migrates as 40-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6647