Learn More
Biomatik Corporation Human Galectin 9 (GAL9) CLIA Kit
Supplier:  Biomatik Corporation EKU09836
Encompass_Preferred
Human Galectin 9 (GAL9) CLIA Kit for detecting GAL9 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500pg/ml. Sensitivity: 0.28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O00182-2. Gene ID: 3965. Target Synonyms: LGALS9; Ecalectin; Lectin,Galactoside-Binding Soluble 9; Tumor antigen HOM-HD-21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4392