Learn More
ACROBiosystems Recombinant Protein;Human Galectin-9 / LGALS9 Protein;HEK293;50UG;LG9,Galectin-9,LGALS9,Ecalectin,Galectin,Gal-9
Supplier: ACROBiosystems LG9H524450UG
Recombinant Protein;Human Galectin-9 / LGALS9 Protein;50UG;Human Galectin-9, His Tag (LG9-H5244) is expressed from human 293 cells (HEK293). It contains AA Ala 2 - Thr 323 (Accession # BAA31542).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE).