Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Galectin-9 Quantikine ELISA Kit
SDP

Catalog No. DGAL90 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DGAL90 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DGAL90 Supplier R&D Systems™ Supplier No. DGAL90
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Galectin-9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Galectin-9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-9 in cell culture supernates (100 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), saliva (100 ul), urine (10 ul), human milk (10 ul).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.028 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human Galectin-9
For Use With (Application) ELISA
Gene ID (Entrez) 3965
Gene Symbol LGALS9
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Saliva (100uL),Urine (10uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Target Galectin-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.