Learn More
Sino Biological Human GALNT10 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 16046H07H50
Encompass_Preferred
A DNA sequence encoding the human GALNT10 (NP_938080.1) (Thr39-Asn603) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-237-3290