Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Gamma Globulin, Purified

Catalog No. MP0855838 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855838 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855838 Supplier MP Biomedicals™ Supplier No. 0855838
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyophilized human gamma globulin with buffer salts

  • Product is the lyophilized powder of human gamma globulin and buffer salts
  • Purified human γ-Globulin can be used as a stabilizer and/or blocker for EIA and RIA test systems
  • Can be used in manufacturing of diagnostic reagents
  • Purified proteins are recommended as ELISA coating antigens and as controls
TRUSTED_SUSTAINABILITY

Specifications

Concentration 17.0 mg/mL Total Protein
Formulation Lyophilized
Quantity 100 mg
Sample Volume 6 mL
Sterility Sterile
Form Lyophilized
Material Powder
Host Species Human
Product Type Lyophilized Powder of Human Gamma Globulin, Buffer Salts

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.