Learn More
TRIBIOSCIENCE INC Human Gamma-H2AX/rH2AX Fast ELISA
SDP
Catalog No. NC3360843
Encompass
Tribioscience’s Fast Human γ-H2AX ELISA is designed to quantitatively detect human γ-H2AX levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 312.5 to 20000 pg/mL.
Catalog No. NC3360843 Supplier TRIBIOSCIENCE INC Supplier No. TBS3265
Tribioscience’s Fast Human γ-H2AX ELISA is designed to quantitatively detect human γ-H2AX levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 312.5 to 20000 pg/mL.