Learn More
Biomatik Corporation Human Gamma-Aminobutyric Acid A Receptor Alpha 2 (gABRa2) ELISA Kit
Supplier:  Biomatik Corporation EKU04331
Encompass_Preferred
Human Gamma-Aminobutyric Acid A Receptor Alpha 2 (gABRa2) ELISA Kit for detecting gABRa2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47869. Gene ID: 2555. Target Synonyms: GABA (A) receptor subunit alpha-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8965