Learn More
Biomatik Corporation Human Gamma-Aminobutyric Acid B Receptor 1 (gABBR1) ELISA Kit
Supplier:  Biomatik Corporation EKU10856
Encompass_Preferred
Human Gamma-Aminobutyric Acid B Receptor 1 (gABBR1) ELISA Kit for detecting gABBR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBS5. Gene ID: 2550. Target Synonyms: GABAB (1e) ; GABABR1; GABA-BR1; GABBR1-3; GPRC3A; HGB1a; Gamma-aminobutyric acid type B receptor subunit 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8277