Learn More
Biomatik Corporation Human Gamma-Glutamyltransferase 5 (gGT5) ELISA Kit
Supplier:  Biomatik Corporation EKU04337
Encompass_Preferred
Human Gamma-Glutamyltransferase 5 (gGT5) ELISA Kit for detecting gGT5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36269. Gene ID: 2687. Target Synonyms: GGTLA1; GGT-REL; Gamma-Glutamyltransferase-Like Activity 1; Gamma-glutamyl transpeptidase-related enzyme; Leukotriene-C4 hydrolase; Glutathione hydrolase 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5655