Learn More
ATCC Human gammaherpesvirus 4 (HHV-4) strain B95-8 (ATCC VR-1492™)
Supplier:  ATCC VR1492
Encompass
Human gammaherpesvirus 4 (HHV-4) strain B95-8 (ATCC VR-1492™), also known as Epstein-Barr virus (EBV), is supplied as 5 x 1 mL vials of virus shed from B95-8 cells. Capable of transforming human B-lymphocytes in vitro, it is used in immunology, oncology, and viral pathogenesis research. Recommended for establishing immortalized lymphoblastoid cell lines.
Recommended companion products:
RPMI-1640 medium (Fisher Cat. No. 50-238-3277, ATCC 30-2001™)
Fetal bovine serum (Fisher Cat. No. 50-238-3771, ATCC 30-2020™).
Catalog No. 50-238-2556