Learn More
BIOHIPPO Human GAP43 Microsample
SDP
Catalog No. 5000313894
Encompass
Human GAP43 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAP43 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GAP43 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GAP43 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GAP43 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13894 Supplier BIOHIPPO Supplier No. BHE15212171
Human GAP43 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAP43 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GAP43 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GAP43 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GAP43 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color