Learn More
Sino Biological Recombinant Human GAP-43 Protein (His Tag)
Supplier:  Sino Biological 12408H08H50
Encompass_Preferred
A DNA sequence encoding the human GAP43 (P17677) (Met 1-Ala 238) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7121