Learn More
Sino Biological Anti-GAP-43 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204154T38100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human GAP43 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3896