Learn More
Sino Biological Anti-GAPDH Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10094MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GAPDH (rh GAPDH; Catalog#10094-H07E; P04406; Met1-Glu335). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0383