Learn More
Raybiotech Inc Human Gas 6 ELISA Kit
Supplier:  Raybiotech Inc ELHGAS61
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.04 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Gas 6.
Catalog No. 50-194-9295