Learn More
Sino Biological Recombinant Human GAS6 Protein (His Tag), HPLC-verified
Catalog No. 502993463
Encompass_Preferred
A DNA sequence encoding the Human GAS6 (NP_000811.1) (Ala49-Ala678) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3463 Supplier Sino Biological Supplier No. 13170H08H21
A DNA sequence encoding the Human GAS6 (NP_000811.1) (Ala49-Ala678) was expressed with a polyhistidine tag at the C-terminus.