Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GASP-1 ELISA Kit

Catalog No. EH209RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH209RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH209RBX10 Supplier Invitrogen™ Supplier No. EH209RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human GASP-1 quantitates human GASP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GASP-1.

Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5JY77
Assay Range 20-5000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; KIAA0443; Per1 interacting protein; per1-interacting protein; Pips2210415K24Rik; 3110031O14Rik; C87852; G protein-coupled receptor associated sorting protein 1; GASP; GASP1; GASP-1; GPRASP1; G-protein coupled receptor-associated sorting protein 1; KIAA0443; Per1 interacting protein; per1-interacting protein; Pips;
Gene ID (Entrez) 9737
Gene Symbol GPRASP1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target GPRASP1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.