Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Gastrin ELISA Kit

Catalog No. EEL058
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL058 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL058 Supplier Invitrogen™ Supplier No. EEL058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Gastrin (GT) ELISA quantitates GT in serum, plasma and other biological fluids Principle of the method: The Competitive Gastrin (GT) ELISA research-use-only kit is designed to quantitatively measure GT in Humans. A GT standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to Human GT is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of Human GT in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01350
Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Gastrin/cholecystokinin
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrinBig gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin;
Gene ID (Entrez) 2520
Gene Symbol Gast
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Gastrin
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.