Learn More
Biomatik Corporation Human Gastrokine 2 (GKN2) ELISA Kit
Supplier:  Biomatik Corporation EKU04352
Encompass_Preferred
Human Gastrokine 2 (GKN2) ELISA Kit for detecting GKN2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86XP6. Gene ID: 200504. Target Synonyms: TFIZ1; BLOT; GDDR; VLTI465; Blottin; Down Regulated In Gastric Cancer; Trefoil factor interactions (z) 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4801