Learn More
Sino Biological Recombinant Anti-GCG/Glucagon Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 110122R060100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the Human GCG/Glucagon.
Catalog No. 50-255-4981