Learn More
Sino Biological Anti-Glucokinase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11078T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Glucokinase / GCK (rh Glucokinase / GCK; Catalog#11078-HNCE; NP_000153.1; Leu2-Gln465). Glucokinase / GCK specific IgG was purified by Human Glucokinase / GCK affinity chromatography.
Catalog No. 50-255-4872