Learn More
Raybiotech Inc Human GDF-11/BMP-11 ELISA Kit
Supplier:  Raybiotech Inc ELHGDF111
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.16 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human GDF-11.
Catalog No. 50-194-9303