Learn More
ACROBiosystems Human GDF-15 200ug
Catalog No. 502535029
Encompass_Preferred
Biotinylated Human GDF-15 Protein HisAvitag (GD5-H81Q3) is expressed from E. coli cells. It contains AA Ala 197 - Ile 308 (Accession # Q99988-1).
Catalog No. 50-253-5029 Supplier ACROBiosystems Supplier No. GD5H81Q3200UG
Biotinylated Human GDF-15 Protein HisAvitag (GD5-H81Q3) is expressed from E. coli cells. It contains AA Ala 197 - Ile 308 (Accession # Q99988-1).