Learn More
R&D Systems™ Human GDF-15 Magnetic Luminex Performance Assay
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Specifications
Specifications
| Product Type | Magnetic Bead-based Assay |
| Quantity | 100 Tests |
| Species | Human |
| Detection Method | Fluorescence |
| Format | 96-well Microplate and Antibody-coated Magnetic Beads |
| For Use With (Application) | Analysis of biochemical parameters |
| For Use With (Equipment) | Luminex MAGPIX Platform |
| Kit Contents | Bead Diluents, Biotin Antibody Diluents, Calibrator Diluents, Foil Plate Sealers, Mixing Bottles, Round-bottom Microplate, Standard Cocktail(s), Streptavidin-PE, Wash Buffer Concentrate |
| Content And Storage | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.