Learn More
Sino Biological Human GDF-15 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786387
Encompass_Preferred
The Human GDF15 ELISA Pair Set is for the quantitative determination of Human GDF15.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human GDF15 coated on a 96-well plate. Standards and samples are added to the wells, and any Human GDF15 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human GDF15 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human GDF15 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86387 Supplier Sino Biological Supplier No. SEK109365
The Human GDF15 ELISA Pair Set is for the quantitative determination of Human GDF15.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human GDF15 coated on a 96-well plate. Standards and samples are added to the wells, and any Human GDF15 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human GDF15 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human GDF15 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.