Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human GDF-15/MIC-1 Recombinant Protein, PeproTech®

Catalog No. 12028C20UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12028C20UG 20 μg
12028C100UG 100 μg
120-28C-1MG 1 mg
12028C250UG 250 μg
12028C500UG 500 μg
120-28C-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12028C20UG Supplier Gibco™ Supplier No. 12028C20UG
  • $224.00
    $190.65 / Each of 1

    Save $33.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GDF-15/MIC-1 is a disulfide linked homodimeric protein consisting of two 112 amino acid polypeptide chains. The calculated molecular weight of Recombinant Human GDF-15/MIC-1 is 24.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99988
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 9518
Molecular Weight (g/mol) 24.6 kDa
Name Human GDF-15/MIC-1
Purification Method Purified
Quantity 20 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Gdf15; GDF-15; growth differentiation factor 15; growth/differentiation factor 15; macrophage inhibiting cytokine-1; macrophage inhibitory cytokine 1; MIC1; MIC-1; NAG1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; PDF; PLAB; Placental bone morphogenetic protein; placental TGF-beta; Prostate differentiation factor; PTGFB; PTGF-beta; SBF
Common Name GDF15
Gene Symbol Gdf15
Biological Activity Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1 osteoblastcells. The expected ED50 for this effect is 75-200 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARNGDHCPLG PGRCCRLHTV RASLEDLGWA DWVLSPREVQ VTMCIGACPS QFRAANMHAQ IKTSLHRLKP DTVPAPCCVP ASYNPMVLIQ KTDTGVSLQT YDDLLAKDCH CI
Content And Storage -20°C
Activity Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1 osteoblastcells. The expected ED50 for this effect is 75-200 ng/ml.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.