Learn More
FUJIFILM BIOSCIENCES INC Human GDF-5 , Recombinant Protein, Shenandoah Biotechnology, 50 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1007150UG
Encompass_Preferred
Growth differentiation factor 5 (GDF-5) is a member of the bone morphogenetic protein (BMP) and transforming growth factor beta (TGF-B) families and functions to regulate cell proliferation and differentiation in embryonic and adult tissues. GDF-5 is expressed in the central nervous system and promotes the survival of dopaminergic neurons in animal models of Parkinson's disease. GDF-5 is also important during chondrogenesis and chondrocyte differentiation.
Catalog No. 50-990-766