Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human GDF-7 Recombinant Protein, PeproTech®

Catalog No. 1203750UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-37-50UG 50 μg
120-37-10UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 120-37-50UG Supplier Gibco™ Supplier No. 1203750UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z4P5
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 151449
Molecular Weight (g/mol) 28 kDa
Name Human GDF-7
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP12; Gdf7; Gdf-7; growth differentiation factor 7; growth/differentiation factor 7
Common Name GDF7
Gene Symbol GDF7
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
Activity Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.