Learn More
Sino Biological Human GDF15 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10936MM41100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GDF15 (rh GDF15; Catalog# 10936-H07Y; NP_004855.2; Ala197-Ile308). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0604