Learn More
Sino Biological Recombinant Human GDNF Protein
Supplier:  Sino Biological 10561HNCH100
Encompass_Preferred
A DNA sequence encoding the mature form of human GDNF (P39905-2) (Arg 83-Ile 185) was expressed and purified, with additional two aa (Gly& Pro) at the N-terminus.
Catalog No. 50-224-6627