Learn More
Gibco™ Human GDNF Recombinant Protein, PeproTech®
Description
The functional human GDNF ligand is a disulfide-linked homodimer consisting of two 15 kDa polypeptide chains called monomers. Each monomer contains seven conserved cysteine residues, including Cys-101, which is used for inter-chain disulfide bridging, and others that are involved in the intramolecular ring formation known as the cysteine-knot configuration. The calculated molecular weight of Recombinant Human GDNF is 30.4 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P39905 |
| For Use With (Application) | Bioactivity, Control, Western Blot Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 2668 |
| Molecular Weight (g/mol) | 30.4 kDa |
| Name | Human GDNF |
| Purification Method | Purified |
| Quantity | 250 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.