Learn More
Abcam Human Gelsolin Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502672417
Encompass_Preferred
Human Gelsolin Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Gelsolin.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2417 Supplier Abcam Supplier No. AB25331210X96TESTS
Human Gelsolin Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Gelsolin.
The product is subject to the following: Abcam Restricted Use Statement