Learn More
Abcam Human Gelsolin ELISA Kit, 10 x 96 Tests
Catalog No. 502656195
Encompass_Preferred
Human Gelsolin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Gelsolin with a sensitivity of 145.98 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6195 Supplier Abcam Supplier No. AB27021510X96TESTS
Add to Cart
Add to Cart
Request a Quote
Human Gelsolin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Gelsolin with a sensitivity of 145.98 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement