Learn More
Biomatik Corporation Human Geminin Coiled Coil Domain Containing Protein 1 (GEMC1) ELISA Kit
Supplier:  Biomatik Corporation EKU12106
Encompass_Preferred
Human Geminin Coiled Coil Domain Containing Protein 1 (GEMC1) ELISA Kit for detecting GEMC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A6NCL1. Gene ID: 647309. Target Synonyms: GMNC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4546