Learn More
Takara Bio HUMAN GENOMIC DNA 100UG
SDP
Catalog No. NC9211717
Encompass
Human Genomic DNA; 100ug; Storage Buffer: 10mM Tris-HCl (7.5 pH), 0.1mM EDTA; Ideal for Southern hybridization and PCR; Suitable for constructing genomic libraries; High molecular weight DNA 50 to 250kb; Storage: 4 deg.C
Catalog No. NC9211717 Supplier Takara Bio Supplier No. 636401
Human Genomic DNA; 100ug; Storage Buffer: 10mM Tris-HCl (7.5 pH), 0.1mM EDTA; Ideal for Southern hybridization and PCR; Suitable for constructing genomic libraries; High molecular weight DNA 50 to 250kb; Storage: 4 deg.C